Assay Details
| Gene Symbol: SEPTIN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599751.3 | XP_005599808.1 | 1077 | 65 | ||
| XM_023618441.1 | XP_023474209.1 | 1363 | 65 | |||
| XM_023618443.1 | XP_023474211.1 | 1363 | 65 | |||
| XM_023618444.1 | XP_023474212.1 | 950 | 65 | |||
| XM_023618445.1 | XP_023474213.1 | 1363 | 65 | |||
More Information
Set Membership: |
|

SEPTIN10
Genomic Map