Assay Details
| Gene Symbol: VSNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503420.5 | XP_001503470.1 | 523 | 86 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

VSNL1| Gene Symbol: VSNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503420.5 | XP_001503470.1 | 523 | 86 | ||