Assay Details
| Gene Symbol: IQCF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005600602.3 | XP_005600659.1 | 206 | 82 | ||
| XM_005600603.3 | XP_005600660.1 | 165 | 82 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005600602.3, XM_005600603.3 transcripts.
Set Membership: |
|

IQCF1
Genomic Map