Assay Details
| Gene Symbol: GJB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005601082.3 | XP_005601139.1 | 1064 | 79 | ||
| XM_005601084.3 | XP_005601141.1 | 1064 | 79 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005601082.3, XM_005601084.3 transcripts.
Set Membership: |
|

GJB6
Genomic Map