Assay Details
| Gene Symbol: SPTSSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005601777.3 | XP_005601834.1 | 2273 | 72 | ||
| XM_014732718.2 | XP_014588204.1 | 2273 | 72 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005601777.3, XM_014732718.2 transcripts.
Set Membership: |
|

SPTSSB
Genomic Map