Assay Details
| Gene Symbol: BSND | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005607023.3 | XP_005607080.1 | 1988 | 58 | ||
| XM_023629202.1 | XP_023484970.1 | 1141 | 58 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: BSND | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005607023.3 | XP_005607080.1 | 1988 | 58 | ||
| XM_023629202.1 | XP_023484970.1 | 1141 | 58 | |||
Set Membership: |
|