Assay Details
| Gene Symbol: HMGN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003364432.3 | XP_003364480.1 | 199 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003364432.3 transcripts.
Set Membership: |
|

HMGN2
Genomic Map