Assay Details
| Gene Symbol: ZKSCAN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014734265.2 | XP_014589751.1 | 174 | 108 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014734265.2 transcripts.
Set Membership: |
|

ZKSCAN8
Genomic Map