Assay Details
| Gene Symbol: ZKSCAN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014734265.2 | XP_014589751.1 | 925 | 80 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZKSCAN8| Gene Symbol: ZKSCAN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014734265.2 | XP_014589751.1 | 925 | 80 | ||
Set Membership: |
|