Assay Details
| Gene Symbol: ZFP57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023624583.1 | XP_023480351.1 | 121 | 108 | ||
| XM_023624584.1 | XP_023480352.1 | 121 | 108 | |||
| XM_023624586.1 | XP_023480354.1 | 122 | 108 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023624583.1, XM_023624584.1, XM_023624586.1 transcripts.
Set Membership: |
|

ZFP57
Genomic Map