Assay Details
| Gene Symbol: ZFP57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023624586.1 | XP_023480354.1 | 165 | 64 | ||
| XM_023624587.1 | XP_023480355.1 | 61 | 64 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023624586.1, XM_023624587.1 transcripts.
Set Membership: |
|

ZFP57
Genomic Map