Assay Details
| Gene Symbol: LOC102151116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005603672.2 | XP_005603729.2 | 228 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005603672.2 transcripts.
Set Membership: |
|

LOC102151116
Genomic Map