Assay Details
| Gene Symbol: YJEFN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001500876.5 | XP_001500926.3 | 888 | 157 | ||
| XM_005604153.3 | XP_005604210.2 | 888 | 157 | |||
More Information
Set Membership: |
|

YJEFN3
Genomic Map