Assay Details
| Gene Symbol: NDUFAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494417.6 | XP_001494467.3 | 248 | 87 | ||
| XM_005604237.3 | XP_005604294.3 | 248 | 87 | |||
| XM_023625639.1 | XP_023481407.1 | 248 | 87 | |||
| XM_023625640.1 | XP_023481408.1 | 248 | 87 | |||
More Information
Set Membership: |
|

NDUFAF2
Genomic Map