Assay Details
| Gene Symbol: OTULINL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023625827.1 | XP_023481595.1 | 431 | 101 | ||
| XM_023625828.1 | XP_023481596.1 | 345 | 101 | |||
| XM_023625829.1 | XP_023481597.1 | 613 | 101 | |||
More Information
Set Membership: |
|

OTULINL
Genomic Map