Assay Details
| Gene Symbol: OTULINL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023625827.1 | XP_023481595.1 | 834 | 135 | ||
| XM_023625828.1 | XP_023481596.1 | 748 | 135 | |||
| XM_023625829.1 | XP_023481597.1 | 1016 | 135 | |||
| GenBank mRNA | JL641073.1 | - | 2 - 3 | 158 | 135 | |
More Information
Set Membership: |
|

OTULINL
Genomic Map