Assay Details
| Gene Symbol: PROKR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014735040.2 | XP_014590526.1 | 190 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014735040.2 transcripts.
Set Membership: |
|

PROKR2
Genomic Map