Assay Details
| Gene Symbol: SIRPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001916116.4 | XP_001916151.2 | 571 | 89 | ||
| XM_005604537.3 | XP_005604594.2 | 572 | 89 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SIRPB2| Gene Symbol: SIRPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001916116.4 | XP_001916151.2 | 571 | 89 | ||
| XM_005604537.3 | XP_005604594.2 | 572 | 89 | |||
Set Membership: |
|