Assay Details
| Gene Symbol: C23H9orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023627673.1 | XP_023483441.1 | 876 | 99 | ||
| GenBank mRNA | JL615905.2 | - | 2 - 3 | 362 | 99 | |
More Information
Set Membership: |
|

C23H9orf64
Genomic Map