Assay Details
| Gene Symbol: DNAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005606645.3 | XP_005606702.1 | 397 | 107 | ||
| XM_023631186.1 | XP_023486954.1 | 771 | 107 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNAL4| Gene Symbol: DNAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005606645.3 | XP_005606702.1 | 397 | 107 | ||
| XM_023631186.1 | XP_023486954.1 | 771 | 107 | |||
Set Membership: |
|