Assay Details
| Gene Symbol: PSKH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498416.4 | XP_001498466.1 | 150 | 90 | ||
| XM_023637163.1 | XP_023492931.1 | 150 | 90 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001498416.4, XM_023637163.1 transcripts.
Set Membership: |
|

PSKH1
Genomic Map