Assay Details
| Gene Symbol: C6H12orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023643931.1 | XP_023499699.1 | 632 | 72 | ||
| XM_023643932.1 | XP_023499700.1 | 633 | 72 | |||
| XM_023643933.1 | XP_023499701.1 | 374 | 72 | |||
| XM_023643934.1 | XP_023499702.1 | 2315 | 72 | |||
| XM_023643935.1 | XP_023499703.1 | 1818 | 72 | |||
| XM_023643936.1 | XP_023499704.1 | 881 | 72 | |||
| XM_023643937.1 | XP_023499705.1 | 778 | 72 | |||
| XM_023643938.1 | XP_023499706.1 | 634 | 72 | |||
| XM_023643939.1 | XP_023499707.1 | 636 | 72 | |||
More Information
Set Membership: |
|

C6H12orf56
Genomic Map