Assay Details
| Gene Symbol: C6H12orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023643931.1 | XP_023499699.1 | 1270 | 103 | ||
| XM_023643932.1 | XP_023499700.1 | 1271 | 103 | |||
| XM_023643933.1 | XP_023499701.1 | 1012 | 103 | |||
| XM_023643934.1 | XP_023499702.1 | 2953 | 103 | |||
| XM_023643935.1 | XP_023499703.1 | 2456 | 103 | |||
| XM_023643936.1 | XP_023499704.1 | 1519 | 103 | |||
| XM_023643937.1 | XP_023499705.1 | 1416 | 103 | |||
| XM_023643938.1 | XP_023499706.1 | 1272 | 103 | |||
| XM_023643939.1 | XP_023499707.1 | 1274 | 103 | |||
More Information
Set Membership: |
|

C6H12orf56
Genomic Map