Assay Details
| Gene Symbol: C6H12orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023643931.1 | XP_023499699.1 | 1584 | 94 | ||
| XM_023643933.1 | XP_023499701.1 | 1326 | 94 | |||
| XM_023643934.1 | XP_023499702.1 | 3267 | 94 | |||
| XM_023643935.1 | XP_023499703.1 | 2770 | 94 | |||
| XM_023643936.1 | XP_023499704.1 | 1833 | 94 | |||
| XM_023643937.1 | XP_023499705.1 | 1730 | 94 | |||
| XM_023643938.1 | XP_023499706.1 | 1586 | 94 | |||

C6H12orf56
Genomic Map