Assay Details
| Gene Symbol: LOC102147893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023632612.1 | XP_023488380.1 | 1636 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023632612.1 transcripts.
Set Membership: |
|

LOC102147893
Genomic Map