Assay Details
| Gene Symbol: TCP10L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005608134.3 | XP_005608191.2 | 1378 | 96 | ||
| XM_005608135.3 | XP_005608192.2 | 1306 | 96 | |||
More Information
Set Membership: |
|

TCP10L2
Genomic Map