Assay Details
| Gene Symbol: STK17A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023638972.1 | XP_023494740.1 | 1061 | 93 | ||
| GenBank mRNA | HQ889903.1 | - | 1 - 2 | 183 | 93 | |
More Information
Set Membership: |
|

STK17A
Genomic Map