Assay Details
| Gene Symbol: TMBIM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005609215.3 | XP_005609272.1 | 691 | 90 | ||
| XM_023639186.1 | XP_023494954.1 | 650 | 90 | |||
| XM_023639187.1 | XP_023494955.1 | 557 | 90 | |||

TMBIM7
Genomic Map