Assay Details
| Gene Symbol: CHRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023639765.1 | XP_023495533.1 | 614 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023639765.1 transcripts.
Set Membership: |
|

CHRM2
Genomic Map