Assay Details
| Gene Symbol: LOC100066339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496641.6 | XP_001496691.4 | 2865 | 129 | ||
| XM_023639791.1 | XP_023495559.1 | 837 | 129 | |||
| XM_023639792.1 | XP_023495560.1 | 839 | 129 | |||
| XM_023639793.1 | XP_023495561.1 | 694 | 129 | |||
| XM_023639794.1 | XP_023495562.1 | 2702 | 129 | |||
| XM_023639795.1 | XP_023495563.1 | 778 | 129 | |||
| XM_023639796.1 | XP_023495564.1 | 662 | 129 | |||
| XM_023639797.1 | XP_023495565.1 | 2939 | 129 | |||
| XM_023639799.1 | XP_023495567.1 | 949 | 129 | |||
| XM_023639800.1 | XP_023495568.1 | 548 | 129 | |||
| XM_023639801.1 | XP_023495569.1 | 696 | 129 | |||
| XM_023639802.1 | XP_023495570.1 | 372 | 129 | |||
| XM_023639803.1 | XP_023495571.1 | 229 | 129 | |||
More Information
Set Membership: |
|

LOC100066339
Genomic Map