Assay Details
| Gene Symbol: LOC100066339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496641.6 | XP_001496691.4 | 3187 | 92 | ||
| XM_023639791.1 | XP_023495559.1 | 1159 | 92 | |||
| XM_023639792.1 | XP_023495560.1 | 1161 | 92 | |||
| XM_023639793.1 | XP_023495561.1 | 1016 | 92 | |||
| XM_023639794.1 | XP_023495562.1 | 3024 | 92 | |||
| XM_023639795.1 | XP_023495563.1 | 1100 | 92 | |||
| XM_023639796.1 | XP_023495564.1 | 984 | 92 | |||
| XM_023639797.1 | XP_023495565.1 | 3261 | 92 | |||
| XM_023639799.1 | XP_023495567.1 | 1271 | 92 | |||
| XM_023639800.1 | XP_023495568.1 | 870 | 92 | |||
| XM_023639801.1 | XP_023495569.1 | 1018 | 92 | |||
| XM_023639802.1 | XP_023495570.1 | 694 | 92 | |||
| XM_023639803.1 | XP_023495571.1 | 551 | 92 | |||
| GenBank mRNA | JL620404.1 | - | 2 - 3 | 207 | 92 | |
More Information
Set Membership: |
|

LOC100066339
Genomic Map