Assay Details
| Gene Symbol: LOC100066339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496641.6 | XP_001496691.4 | 3312 | 89 | ||
| XM_023639791.1 | XP_023495559.1 | 1284 | 89 | |||
| XM_023639793.1 | XP_023495561.1 | 1141 | 89 | |||
| XM_023639794.1 | XP_023495562.1 | 3149 | 89 | |||
| XM_023639795.1 | XP_023495563.1 | 1225 | 89 | |||
| XM_023639796.1 | XP_023495564.1 | 1109 | 89 | |||
| XM_023639797.1 | XP_023495565.1 | 3386 | 89 | |||
| XM_023639799.1 | XP_023495567.1 | 1396 | 89 | |||
| XM_023639800.1 | XP_023495568.1 | 995 | 89 | |||
| XM_023639802.1 | XP_023495570.1 | 819 | 89 | |||
| XM_023639803.1 | XP_023495571.1 | 676 | 89 | |||
| GenBank mRNA | JL620404.1 | - | 4 - 5 | 332 | 89 | |
More Information
Set Membership: |
|

LOC100066339
Genomic Map