Assay Details
| Gene Symbol: MRPS33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498241.5 | XP_001498291.1 | 150 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001498241.5 transcripts.
Set Membership: |
|

MRPS33
Genomic Map