Assay Details
| Gene Symbol: LOC100062440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005612783.3 | XP_005612840.2 | 313 | 139 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005612783.3 transcripts.
Set Membership: |
|

LOC100062440
Genomic Map