Assay Details
| Gene Symbol: ZNF532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023647888.1 | XP_023503656.1 | 789 | 135 | ||
| XM_023647889.1 | XP_023503657.1 | 295 | 135 | |||
| XM_023647890.1 | XP_023503658.1 | 231 | 135 | |||
| XM_023647893.1 | XP_023503661.1 | 789 | 135 | |||
| XM_023647894.1 | XP_023503662.1 | 788 | 135 | |||
| XM_023647895.1 | XP_023503663.1 | 788 | 135 | |||
| XM_023647897.1 | XP_023503665.1 | 788 | 135 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023647888.1, XM_023647889.1, XM_023647890.1, XM_023647893.1, XM_023647894.1, XM_023647895.1, XM_023647897.1 transcripts.
Set Membership: |
|

ZNF532
Genomic Map