Assay Details
| Gene Symbol: ZNF532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023647888.1 | XP_023503656.1 | 4267 | 78 | ||
| XM_023647889.1 | XP_023503657.1 | 3773 | 78 | |||
| XM_023647890.1 | XP_023503658.1 | 3709 | 78 | |||
| XM_023647891.1 | XP_023503659.1 | 3868 | 78 | |||
| XM_023647892.1 | XP_023503660.1 | 3691 | 78 | |||
| XM_023647894.1 | XP_023503662.1 | 4176 | 78 | |||

ZNF532
Genomic Map