Assay Details
| Gene Symbol: RAB9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005613989.3 | XP_005614046.1 | 280 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005613989.3 transcripts.
Set Membership: |
|

RAB9A
Genomic Map