Assay Details
| Gene Symbol: CXHXorf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023633577.1 | XP_023489345.1 | 1014 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CXHXorf38| Gene Symbol: CXHXorf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023633577.1 | XP_023489345.1 | 1014 | 67 | ||
Set Membership: |
|