Assay Details
| Gene Symbol: GPR174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005614296.3 | XP_005614353.1 | 135 | 100 | ||
| XM_005614297.3 | XP_005614354.1 | 135 | 100 | |||
| XM_005614298.3 | XP_005614355.1 | 135 | 100 | |||
| XM_023633326.1 | XP_023489094.1 | 135 | 100 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005614296.3, XM_005614297.3, XM_005614298.3, XM_023633326.1 transcripts.
Set Membership: |
|

GPR174
Genomic Map