Assay Details
| Gene Symbol: GPR174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005614296.3 | XP_005614353.1 | 266 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005614296.3 transcripts.
Set Membership: |
|

GPR174
Genomic Map