Assay Details
| Gene Symbol: SEPTIN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014729135.2 | XP_014584621.1 | 1287 | 101 | ||
| XM_014729136.2 | XP_014584622.1 | 1168 | 101 | |||
| XM_023634069.1 | XP_023489837.1 | 1285 | 101 | |||
| XM_023634070.1 | XP_023489838.1 | 1285 | 101 | |||
| XM_023634071.1 | XP_023489839.1 | 1285 | 101 | |||
| XM_023634072.1 | XP_023489840.1 | 1284 | 101 | |||
| XM_023634073.1 | XP_023489841.1 | 1283 | 101 | |||
| XM_023634074.1 | XP_023489842.1 | 1283 | 101 | |||
| XM_023634075.1 | XP_023489843.1 | 1281 | 101 | |||
| XM_023634076.1 | XP_023489844.1 | 1282 | 101 | |||
| XM_023634077.1 | XP_023489845.1 | 1282 | 101 | |||
More Information
Set Membership: |
|

SEPTIN6
Genomic Map