Assay Details
| Gene Symbol: PRRG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005614606.3 | XP_005614663.1 | 301 | 83 | ||
| XM_014729261.2 | XP_014584747.1 | 325 | 83 | |||
| XM_023633235.1 | XP_023489003.1 | 477 | 83 | |||
| XM_023633236.1 | XP_023489004.1 | 2340 | 83 | |||
| XM_023633237.1 | XP_023489005.1 | 451 | 83 | |||
| XM_023633238.1 | XP_023489006.1 | 290 | 83 | |||
| XM_023633239.1 | XP_023489007.1 | 2309 | 83 | |||
More Information
Set Membership: |
|

PRRG3
Genomic Map