Assay Details
| Gene Symbol: OTUD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023632358.1 | XP_023488126.1 | 1515 | 79 | ||
| XM_023632360.1 | XP_023488128.1 | 1514 | 79 | |||
| XM_023632361.1 | XP_023488129.1 | 1515 | 79 | |||
| GenBank mRNA | JL622144.1 | - | 1 | 123 | 79 | |
More Information
Set Membership: |
|

OTUD1
Genomic Map