Assay Details
| Gene Symbol: EEF1AKMT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001490082.6 | XP_001490132.1 | 1986 | 84 | ||
| GenBank mRNA | JL629439.1 | - | 1 | 1016 | 84 | |
More Information
Set Membership: |
|

EEF1AKMT3
Genomic Map