Assay Details
| Gene Symbol: INSL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023625458.1 | XP_023481226.1 | 224 | 103 | ||
| GenBank mRNA | AB033169.1 | - | 1 - 2 | 92 | 103 | |
More Information
Set Membership: |
|

INSL3
Genomic Map