Assay Details
| Gene Symbol: KCTD16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599333.3 | XP_005599390.1 | 1528 | 109 | ||
| XM_005599334.3 | XP_005599391.1 | 1421 | 109 | |||
| XM_014730599.2 | XP_014586085.1 | 1656 | 109 | |||
More Information
Set Membership: |
|

KCTD16
Genomic Map