Assay Details
| Gene Symbol: SERTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023621516.1 | XP_023477284.1 | 3116 | 108 | ||
| XM_023621517.1 | XP_023477285.1 | 4563 | 108 | |||
More Information
Set Membership: |
|

SERTM1
Genomic Map