Assay Details
| Gene Symbol: ZFP69B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005607170.3 | XP_005607227.1 | 3176 | 85 | ||
| XM_005607172.3 | XP_005607229.1 | 3022 | 85 | |||
More Information
Set Membership: |
|

ZFP69B
Genomic Map