Assay Details
| Gene Symbol: TIGD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023636428.1 | XP_023492196.1 | 2307 | 59 | ||
| XM_023636429.1 | XP_023492197.1 | 2307 | 59 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023636428.1, XM_023636429.1 transcripts.
Set Membership: |
|

TIGD4
Genomic Map