Assay Details
| Gene Symbol: ASCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023631353.1 | XP_023487121.1 | 478 | 133 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023631353.1 transcripts.
Set Membership: |
|

ASCL1
Genomic Map